missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorododecane 97.0+%, TCI America™

Catalog No. C0163500ML
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0163500ML 500 mL
C016325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. C0163500ML missing translation for 'mfr' TCI America missing translation for 'supplierNo' C0163500ML

Chemical Identifiers

CAS 112-52-7
Molecular Formula C12H25Cl
Molecular Weight (g/mol) 204.78
MDL Number MFCD00013688
InChI Key YAYNEUUHHLGGAH-UHFFFAOYSA-N
Synonym lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk
PubChem CID 8192
IUPAC Name 1-chlorododecane
SMILES CCCCCCCCCCCCCl

Specifications

Color Colorless
Boiling Point 260°C
Quantity 500 mL
UN Number 3082
Formula Weight 204.78
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorododecane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes