missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -123°C |
| Color | Incolore |
| Boiling Point | 78°C |
| Quantity | 25 mL |
| UN Number | 1127 |
| Formula Weight | 92.57 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 1-Chlorobutane |
Sécurité et manipulation
| EINECSNumber | (2)-0060 |
| RTECSNumber | EJ6300000 |
| TSCA | Yes |