missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-9,10-bis(phenylethynyl)anthracene 96.0+%, TCI America™

Catalog No. C12891G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12891G 1 g
C128910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12891G Supplier TCI America Supplier No. C12891G
Only null left

Chemical Identifiers

CAS 41105-35-5
Molecular Formula C30H17Cl
Molecular Weight (g/mol) 412.92
MDL Number MFCD00012047
InChI Key IMMCAKJISYGPDQ-UHFFFAOYSA-N
Synonym 1-chloro-9,10-bis phenylethynyl anthracene, 1-chloro-bpea, anthracene, 1-chloro-9,10-bis phenylethynyl, 1-chloro-9,10-bis 2-phenylethynyl anthracene, anthracene, 9,10-bis phenylethynyl-1-chloro, anthracene, 1-chloro-9,10-bis 2-phenylethynyl, 1-chloro-bpee, cbpea, 1-cl-bpee, acmc-1an87
PubChem CID 170465
IUPAC Name 1-chloro-9,10-bis(2-phenylethynyl)anthracene
SMILES ClC1=C2C(=CC=C1)C(C#CC1=CC=CC=C1)=C1C=CC=CC1=C2C#CC1=CC=CC=C1

Specifications

Melting Point 203°C
Quantity 1 g
Formula Weight 412.92
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-9,10-bis(phenylethynyl)anthracene