missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodopropane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. C10375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10375G 5 g
C103725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10375G Supplier TCI America Supplier No. C10375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6940-76-7
Molecular Formula C3H6ClI
Molecular Weight (g/mol) 204.435
MDL Number MFCD00001096
InChI Key SFOYQZYQTQDRIY-UHFFFAOYSA-N
Synonym propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane
PubChem CID 81363
IUPAC Name 1-chloro-3-iodopropane
SMILES C(CCl)CI

Specifications

Color Yellow
Boiling Point 171°C
Quantity 5 g
Formula Weight 204.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-iodopropane (stabilized with Copper chip)