missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodopropane, 98%, stab. with copper

Catalog No. AAA1348936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348936 500 g
AAA1348914 25 g
AAA1348922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1348936 Supplier Thermo Scientific Chemicals Supplier No. A1348936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6940-76-7 | C3H6ClI | 204.435 g/mol

1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 6940-76-7
Molecular Formula C3H6ClI
Molecular Weight (g/mol) 204.435
MDL Number MFCD00001096
InChI Key SFOYQZYQTQDRIY-UHFFFAOYSA-N
Synonym propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane
PubChem CID 81363
IUPAC Name 1-chloro-3-iodopropane
SMILES C(CCl)CI

Specifications

Density 1.904
Boiling Point 170°C to 172°C
Flash Point >110°C (230°F)
Refractive Index 1.5463
Quantity 500 g
Beilstein 1731115
Sensitivity Light sensitive
Solubility Information Difficult to mix.
Formula Weight 204.44
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-iodopropane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-088-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only