missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodobenzene, 98%

Catalog No. AAA1556722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1556722 100 g
AAA1556706 5 g
AAA1556714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1556722 Supplier Thermo Scientific Chemicals Supplier No. A1556722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 625-99-0 | C6H4ClI | 238.45 g/mol

1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 67.2 mg/L.

Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 625-99-0
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001046
InChI Key JMLWXCJXOYDXRN-UHFFFAOYSA-N
Synonym 3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
PubChem CID 12269
IUPAC Name 1-chloro-3-iodobenzene
SMILES ClC1=CC=CC(I)=C1

Specifications

Density 1.927
Boiling Point 228°C
Flash Point 102°C (216°F)
Refractive Index 1.631
Quantity 100 g
Beilstein 1904539
Sensitivity Light sensitive
Solubility Information Soluble in water 67.2mg/L.
Formula Weight 238.46
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-iodobenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-920-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only