missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodobenzene (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. C018925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C018925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C018925G Supplier TCI America Supplier No. C018925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzene
SMILES ClC1=CC=CC=C1I

Specifications

Melting Point 1°C
Color Yellow
Boiling Point 232°C
Quantity 25 g
Formula Weight 238.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-iodobenzene (stabilized with Copper chip)