missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodobenzène, 99%

Numéro de catalogue. AC249460050
Modifier l'affichage
Click to view available options
Quantity:
5 g
50 g
Packaging:
Bouteille en verre
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC249460050 5 g Bouteille en verre
AC249460500 50 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC249460050 Fournisseur Thermo Scientific Chemicals Code fournisseur 249460050
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

CAS : 615-41-8 | C6H4ClI | 238,45 g/mol

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Identifiants chimiques

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzène
SMILES ClC1=CC=CC=C1I

Spécifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.0°C
Color Jaune
Density 1.9500g/mL
Boiling Point 234.0°C to 235.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Bouteille en verre
Refractive Index 1.6330 to 1.6350
Linear Formula ClC6H4I
Quantity 5 g
Beilstein 05, 220
Specific Gravity 1.95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 238.46
Percent Purity 99%
Physical Form Liquide
Chemical Name or Material 1-Chloro-2-iodobenzene, 99%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-423-4
TSCA TSCA

Utilisation uniquement de la recherche RUO –