missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-fluorobenzene 98.0+%, TCI America™

Catalog No. p-7124543
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C064710G 10 g
C064725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C064710G Supplier TCI America Supplier No. C064710G

Chemical Identifiers

CAS 348-51-6
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000533
InChI Key ZCJAYDKWZAWMPR-UHFFFAOYSA-N
Synonym 2-chlorofluorobenzene, o-chlorofluorobenzene, benzene, 1-chloro-2-fluoro, o-fluorochlorobenzene, 1-fluoro-2-chlorobenzene, 1-chloro-2-fluoro-benzene, benzene, chlorofluoro, 2-chloro-1-fluorobenzene, chlorofluorobenzene, 2-fluorochlorobenzene
PubChem CID 9583
IUPAC Name 1-chloro-2-fluorobenzene
SMILES FC1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 138°C
Quantity 10 g
UN Number 1993
Formula Weight 130.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-fluorobenzene