missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Iodide 96.0+%, TCI America™

Catalog No. B27085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27085G 5 g
B270825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27085G Supplier TCI America Supplier No. B27085G
Only null left

Chemical Identifiers

CAS 65039-05-6
Molecular Formula C8H15IN2
Molecular Weight (g/mol) 266.126
InChI Key XREPTGNZZKNFQZ-UHFFFAOYSA-M
PubChem CID 11448496
ChEBI CHEBI:61335
IUPAC Name 1-butyl-3-methylimidazol-3-ium;iodide
SMILES CCCCN1C=C[N+](=C1)C.[I-]

Specifications

Quantity 5 g
Formula Weight 266.13
Percent Purity ≥96.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Iodide