missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Bromide 98.0+%, TCI America™

Catalog No. B21935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21935G Supplier TCI America Supplier No. B21935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85100-77-2
Molecular Formula C8H15BrN2
Molecular Weight (g/mol) 219.126
MDL Number MFCD03427611
InChI Key KYCQOKLOSUBEJK-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium bromide, 3-butyl-1-methyl-1h-imidazol-3-ium bromide, bmimbr, 1-n-butyl-3-methylimidazolium bromide, 1-butyl-3-methyl-1h-imidazol-3-ium bromide, bmim br, c4mim bromide, ksc491c7d, 1-butyl-3-methylimidazoliumbromide
PubChem CID 2734236
IUPAC Name 1-butyl-3-methylimidazol-3-ium;bromide
SMILES CCCCN1C=C[N+](=C1)C.[Br-]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 219.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Butyl-3-methylimidazolium Bromide