missing translation for 'onlineSavingsMsg'
Learn More

1-Butoxy-4-eth-1-ynylbenzene, 97%, Thermo Scientific™

Catalog No. p-2695655
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB09896DA 1g Amber Glass Bottle
BTB09896EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB09896DA Supplier Thermo Scientific Chemicals Supplier No. BTB09896DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79887-15-3
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00173878
InChI Key JMLXBCSKFMMFGF-UHFFFAOYSA-N
Synonym 1-butoxy-4-eth-1-ynylbenzene, benzene,1-butoxy-4-ethynyl, 4-n-butoxyphenylacetylene, 4-butoxyphenylacetylene, 1-ethynyl-4-butoxybenzene, 1-butoxy-4-ethynyl-benzene
PubChem CID 2775120
IUPAC Name 1-butoxy-4-ethynylbenzene
SMILES CCCCOC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)3OC6H4Cidentical toCH
Quantity 1g
Formula Weight 174.24
Percent Purity 97%
Chemical Name or Material 1-Butoxy-4-eth-1-ynylbenzene