missing translation for 'onlineSavingsMsg'
Learn More

1-Butene (ca. 10% in Toluene), TCI America™

Catalog No. B4411100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4411100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4411100ML Supplier TCI America Supplier No. B4411100ML
Only null left

Chemical Identifiers

CAS 106-98-9
Molecular Formula C4H8
Molecular Weight (g/mol) 56.108
MDL Number MFCD00009383
InChI Key VXNZUUAINFGPBY-UHFFFAOYSA-N
Synonym 1-butene, ethylethylene, 1-butylene, butene, butene-1, alpha-butylene, alpha-butene, butylene, n-butylene, n-butene
PubChem CID 7844
ChEBI CHEBI:48362
IUPAC Name but-1-ene
SMILES CCC=C

Specifications

Color Yellow
Quantity 100 mL
UN Number 1993
Formula Weight 56.11
Physical Form Liquid
Chemical Name or Material 1-Butene (ca. 10% in Toluene)
EINECSNumber (2)-0016
RTECSNumber EM2920500
TSCA Yes
Recommended Storage Refrigerator