missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol 99.0+%, TCI America™

Catalog No. B0704500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0704500ML 500 mL
B070425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0704500ML Supplier TCI America Supplier No. B0704500ML
Only null left

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym Butyl Alcohol
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 1120
Boiling Point 118°C
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1-Butanol
EINECSNumber (2)-3049
RTECSNumber EO1400000
TSCA Yes