missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol (HPLC), Fisher Chemical™

Catalog No. A383SK4
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
EcoSafPak™ Amber Glass Bottle
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A383SK4 4 L Safe-Cote™ Amber Glass 
A383SK1 1 L Safe-Cote™ Amber Glass 
A3831 1 L Wide Neck Amber Bottle
A3834 4 L EcoSafPak™ Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A383SK4 Supplier Thermo Fisher Scientific Supplier No. FLA383SK4

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89°C
Absorbance 1.00 AU max. at 215nm, .50 AU max. at 220nm, 0.20 AU max. at 230nm, 0.025 AU max. at 254nm
Color Colorless
Density 0.810g/cm³
Quantity 4 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.3952 to 1.3992 (at 25°C)
Residue after Evaporation 0.005% max.
Assay Percent Range ≥99.4 %
Titratable Acid 0.0008mEq/g max.
Vapor Pressure 6.7mbar at 20°C
Linear Formula CH3CH2CH2CH2OH
Viscosity 2.95 mPa/s at 20°C
UN Number 1120; Colorless
Water 0.1% max.
Boiling Point 117.6°C
Formula Weight 74.12
Percent Purity ≥99.4%
Grade Also meets ACS specifications
Chemical Name or Material 1-Butanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid