missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol 99.0+%, TCI America™

Catalog No. B0228100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0228100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0228100ML Supplier TCI America Supplier No. B0228100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Color Colorless
Quantity 100 mL
Physical Form Clear Liquid at 20°C
UN Number 1120
Boiling Point 118°C
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1-Butanol [for Spectrophotometry]
EINECSNumber (2)-3049
RTECSNumber EO1400000
TSCA Yes