missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoanthracene 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 100°C |
| Quantity | 500 mg |
| Formula Weight | 257.13 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Bromoanthracene |
Safety and Handling
| TSCA | No |