missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-methylpentane 98.0+%, TCI America™
Supplier: TCI America B101725ML
Specifications
| 1-Bromo-4-methylpentane | |
| Colorless | |
| C6H13Br | |
| 25 mL | |
| 4-Methylpentyl Bromide | |
| CC(C)CCCBr | |
| 165.074 | |
| 165.07 | |
| Liquid |
| 626-88-0 | |
| 148°C | |
| MFCD00013544 | |
| 1993 | |
| XZKFBZOAIGFZSU-UHFFFAOYSA-N | |
| 1-bromo-4-methylpentane | |
| 69385 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 626-88-0 | |
| 165.074 | |
| XZKFBZOAIGFZSU-UHFFFAOYSA-N | |
| 69385 | |
| CC(C)CCCBr |
| C6H13Br | |
| MFCD00013544 | |
| 4-Methylpentyl Bromide | |
| 1-bromo-4-methylpentane |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes