accessibility menu, dialog, popup

UserName

CAS RN 626-88-0

H3CCH3Br

CAS RN 626-88-0

IUPAC Name: 1-bromo-4-methylpentane
Synonyms: pentane, 1-bromo-4-methyl-
Molecular Weight (g/mol): 165.07
Molecular Formula: C6H13Br
InChi Key: XZKFBZOAIGFZSU-UHFFFAOYSA-N
SMILES: CC(C)CCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results