missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-methylpentane 98.0+%, TCI America™

Catalog No. p-7118806
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10175ML 5 mL
B101725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10175ML Supplier TCI America Supplier No. B10175ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-88-0
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.074
MDL Number MFCD00013544
InChI Key XZKFBZOAIGFZSU-UHFFFAOYSA-N
Synonym 4-Methylpentyl Bromide
PubChem CID 69385
IUPAC Name 1-bromo-4-methylpentane
SMILES CC(C)CCCBr

Specifications

Color Colorless
Boiling Point 148°C
Quantity 5 mL
UN Number 1993
Formula Weight 165.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-methylpentane
EINECSNumber (2)-0067
TSCA Yes