missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-hexylbenzene 90.0+%, TCI America™

Catalog No. B16065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16065G 5 g
B160625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16065G Supplier TCI America Supplier No. B16065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23703-22-2
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.17
MDL Number MFCD00061114
InChI Key MXHOLIARBWJKCR-UHFFFAOYSA-N
Synonym 1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
PubChem CID 598186
IUPAC Name 1-bromo-4-hexylbenzene
SMILES CCCCCCC1=CC=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 241.17
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-hexylbenzene