missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoronaphthalene, 98%

Catalog No. AAA1693414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1693414 25 g
AAA1693403 1 g
AAA1693406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1693414 Supplier Thermo Scientific Chemicals Supplier No. A1693414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 341-41-3 | C10H6BrF | 225.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 341-41-3
Molecular Formula C10H6BrF
Molecular Weight (g/mol) 225.06
MDL Number MFCD00051473
InChI Key VAUJZKBFENPOCH-UHFFFAOYSA-N
Synonym 4-bromo-1-fluoronaphthalene, 1-fluoro-4-bromonaphthalene, naphthalene, 1-bromo-4-fluoro, acmc-1cnye, 1-bromo 4-fluoronaphthalene, 1-bromo-4-fluoro naphthalene, 1-bromo-4-fluoro-naphthalene, ksc493c4t, 1-bromo-4-fluoronaphthalene
PubChem CID 67647
IUPAC Name 1-bromo-4-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)F

Specifications

Melting Point 33°C to 37°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2085804
Formula Weight 225.06
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-fluoronaphthalene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-434-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only