missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluorobutane 98.0+%, TCI America™

Catalog No. B53315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53315G 5 g
B533125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53315G Supplier TCI America Supplier No. B53315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 462-72-6
Molecular Formula C4H8BrF
Molecular Weight (g/mol) 155.01
MDL Number MFCD00209551
InChI Key WMCUHRDQSHQNRW-UHFFFAOYSA-N
PubChem CID 10022
IUPAC Name 1-bromo-4-fluorobutane
SMILES C(CCBr)CF

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 1993
Synonym 4-fluorobutyl bromide, butane, 1-bromo-4-fluoro, butane,1-bromo-4-fluoro, fluorobutyl bromide, acmc-20aoww, 4-bromo-1-fluorobutane, 1-bromo-4-fluoro-butane, 4-01-00-00263 beilstein handbook reference, 1-bromo-4-fluorobutane, 1-bromo-4-fluoro-butan;4-fluoro-1-bromobutane
Formula Weight 155.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-fluorobutane