missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-ethynylbenzene 98.0+%, TCI America™

Catalog No. B37015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37015G 5 g
B37011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37015G Supplier TCI America Supplier No. B37015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-96-1
Molecular Formula C8H5Br
Molecular Weight (g/mol) 181.032
MDL Number MFCD00168822
InChI Key LTLVZQZDXQWLHU-UHFFFAOYSA-N
Synonym 4-bromophenylacetylene, benzene, 1-bromo-4-ethynyl, 1-bromo-4-ethynyl-benzene, 4-bromophenyl acetylene, 4'-bromophenyl acetylene, 4'-bromophenylacetylene, p-bromophenylacetylene, p-bromophenyl acetylene, 4-bromphenyl acetylene, 4-bromo-1-ethynylbenzene
PubChem CID 136603
IUPAC Name 1-bromo-4-ethynylbenzene
SMILES C#CC1=CC=C(C=C1)Br

Specifications

Melting Point 66°C
Color Yellow
Boiling Point 92°C
Quantity 5 g
UN Number 3077
Formula Weight 181.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-ethynylbenzene