missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-nitrobenzene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 55°C |
| Color | Yellow |
| Boiling Point | 257°C |
| Quantity | 500 g |
| UN Number | 3459 |
| Formula Weight | 202.01 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Bromo-3-nitrobenzene |
Safety and Handling
| RTECSNumber | CY9040500 |
| TSCA | Yes |