missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-iodobenzene (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. B12365G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12365G 5 g
B123625G 25 g
B1236100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B12365G Supplier TCI America Supplier No. B12365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 591-18-4
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.91
MDL Number MFCD00001043
InChI Key CTPUUDQIXKUAMO-UHFFFAOYSA-N
Synonym 3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
PubChem CID 11561
IUPAC Name 1-bromo-3-iodobenzene
SMILES BrC1=CC=CC(I)=C1

Specifications

Melting Point -9°C
Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 282.91
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-iodobenzene (stabilized with Copper chip)