missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluorobenzene, 99%

Catalog No. AAA1085830
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1085830 250 g
AAA1085814 25 g
AAA1085818 50 g
AAA1085822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1085830 Supplier Thermo Scientific Chemicals Supplier No. A1085830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1073-06-9 | C6H4BrF | 175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073-06-9
Molecular Formula C6H4BrF
MDL Number MFCD00000326
InChI Key QDFKKJYEIFBEFC-UHFFFAOYSA-N
Synonym 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
PubChem CID 14082
IUPAC Name 1-bromo-3-fluorobenzene

Specifications

Melting Point -8°C
Density 1.594
Boiling Point 148°C to 150°C
Flash Point 46°C (115°F)
Odor Aromatic
Refractive Index 1.5275
Quantity 250 g
UN Number UN1993
Beilstein 1853969
SMILES C1=CC(=CC(=C1)Br)F
Molecular Weight (g/mol) 175
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-fluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 214-023-0
RTECSNumber DA1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only