missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,7-dimethyloctane 93.0+%, TCI America™
Supplier: TCI America B440125G
Specifications
| 1-Bromo-3,7-dimethyloctane | |
| Yellow | |
| C10H21Br | |
| 25 g | |
| VGSUDZKDSKCYJP-UHFFFAOYSA-N | |
| 1-bromo-3,7-dimethyloctane | |
| 137914 | |
| ≥93.0% (GC) |
| 3383-83-3 | |
| 217°C | |
| MFCD00039201 | |
| 3,7-Dimethyloctyl Bromide | |
| CC(C)CCCC(C)CCBr | |
| 221.182 | |
| 221.18 | |
| Liquid |
Chemical Identifiers
| 3383-83-3 | |
| 221.182 | |
| VGSUDZKDSKCYJP-UHFFFAOYSA-N | |
| 137914 | |
| CC(C)CCCC(C)CCBr |
| C10H21Br | |
| MFCD00039201 | |
| 3,7-Dimethyloctyl Bromide | |
| 1-bromo-3,7-dimethyloctane |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : No