missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,7-dimethyloctane 93.0+%, TCI America™

Catalog No. p-7123517
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44015G 5 g
B440125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44015G Supplier TCI America Supplier No. B44015G

Chemical Identifiers

CAS 3383-83-3
Molecular Formula C10H21Br
Molecular Weight (g/mol) 221.182
MDL Number MFCD00039201
InChI Key VGSUDZKDSKCYJP-UHFFFAOYSA-N
Synonym 3,7-Dimethyloctyl Bromide
PubChem CID 137914
IUPAC Name 1-bromo-3,7-dimethyloctane
SMILES CC(C)CCCC(C)CCBr

Specifications

Color Yellow
Boiling Point 217°C
Quantity 5 g
Formula Weight 221.18
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,7-dimethyloctane
EINECSNumber (2)-0067
TSCA No