CAS RN 3383-83-3
IUPAC Name:
1-bromo-3,7-dimethyloctane
Synonyms:
1-bromo-3,7-dimethyloctane
Molecular Weight (g/mol):
221.18
Molecular Formula:
C10H21Br
InChi Key:
VGSUDZKDSKCYJP-UHFFFAOYNA-N
SMILES:
CC(C)CCCC(C)CCBr
1
–
1
of
1
results
1
1-Bromo-3,7-dimethyloctane 93.0+%, TCI America™
CAS: 3383-83-3 Molecular Formula: C10H21Br Molecular Weight (g/mol): 221.182 MDL Number: MFCD00039201 InChI Key: VGSUDZKDSKCYJP-UHFFFAOYSA-N Synonym: 3,7-Dimethyloctyl Bromide PubChem CID: 137914 IUPAC Name: 1-bromo-3,7-dimethyloctane SMILES: CC(C)CCCC(C)CCBr