missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dinitrobenzene, 98%

Catalog No. AAH6677406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6677406 5 g
AAH6677403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6677406 Supplier Thermo Scientific Chemicals Supplier No. H6677406

CAS: 18242-39-2 | C6H3BrN2O4 | 247.004 g/mol

It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 18242-39-2
Molecular Formula C6H3BrN2O4
Molecular Weight (g/mol) 247.004
MDL Number MFCD00156596
InChI Key OLDMYNWXIGPOCI-UHFFFAOYSA-N
PubChem CID 44534
IUPAC Name 1-bromo-3,5-dinitrobenzene
SMILES C1=C(C=C(C=C1[N+](=O)[O-])Br)[N+](=O)[O-]

Specifications

Color White
Quantity 5 g
UN Number UN3459
Beilstein 2053146
Synonym 1-bromo-3,5-dinitro-benzene, benzene, 1-bromo-3,5-dinitro, benzene,1-bromo-3,5-dinitro, 3,5-dinitrobromobenzene, 1-bromo-3,5-dinitro benzene, 5-bromo-1,3-dinitrobenzene, bromodinitrobenzene, pubchem21414, acmc-1b5tp, 3-bromo 1,5-dinitrobenzene
Solubility Information Slightly soluble in water.
Formula Weight 247
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Bromo-3,5-dinitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only