missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4-dichlorobenzene 98.0+%, TCI America™

Catalog No. B0983500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0983500G 500 g
B098325G 25 g
B0983100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0983500G Supplier TCI America Supplier No. B0983500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18282-59-2
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00040849
InChI Key CFPZDVAZISWERM-UHFFFAOYSA-N
Synonym 1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
PubChem CID 29013
IUPAC Name 4-bromo-1,2-dichlorobenzene
SMILES ClC1=CC=C(Br)C=C1Cl

Specifications

Color White-Yellow
Boiling Point 124°C
Quantity 500 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Lumps
Chemical Name or Material 1-Bromo-3,4-dichlorobenzene