missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-nitrobenzene 99.0+%, TCI America™

Catalog No. B0622500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0622500G 500 g
B062225G 25 g
B0622100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0622500G Supplier TCI America Supplier No. B0622500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 577-19-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD00007045
InChI Key ORPVVAKYSXQCJI-UHFFFAOYSA-N
Synonym 2-bromonitrobenzene, o-bromonitrobenzene, o-nitrobromobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, o-nitrophenyl bromide, 2-bromo-1-nitrobenzene, 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, benzene, bromonitro
PubChem CID 11341
IUPAC Name 1-bromo-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 41°C
Boiling Point 264°C
Quantity 500 g
UN Number 3459
Formula Weight 202.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-nitrobenzene
RTECSNumber CY9040450
TSCA Yes
Recommended Storage Refrigerator