missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-isopropoxybenzene 97.0+%, TCI America™

Catalog No. B31005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31005G 5 g
B310025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31005G Supplier TCI America Supplier No. B31005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 701-07-5
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00070759
InChI Key MMORVPBHAHXAHH-UHFFFAOYSA-N
Synonym 1-bromo-2-isopropoxybenzene, 2-2'-bromophenoxy propane, 1-bromo-2-propan-2-yloxy benzene, 1-bromo-2-isopropoxy benzene, 2-2-bromophenoxy propane, o-bromoisopropoxybenzene, 2-bromoisopropoxybenzene, 1-bromo-2-1-methylethoxy benzene, 2-bromophenyl isopropyl ether, pubchem3828
PubChem CID 21925427
IUPAC Name 1-bromo-2-propan-2-yloxybenzene
SMILES CC(C)OC1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 73°C
Quantity 5 g
Formula Weight 215.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-isopropoxybenzene
EINECSNumber (3)-0845
TSCA No