missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-isopropoxybenzene, 97%

Catalog No. AAB2568806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2568806 5 g
AAB2568814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2568806 Supplier Thermo Scientific Chemicals Supplier No. B2568806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 701-07-5 | C9H11BrO | 215.09 g/mol

1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Chemical Identifiers

CAS 701-07-5
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00070759
InChI Key MMORVPBHAHXAHH-UHFFFAOYSA-N
Synonym 1-bromo-2-isopropoxybenzene, 2-2'-bromophenoxy propane, 1-bromo-2-propan-2-yloxy benzene, 1-bromo-2-isopropoxy benzene, 2-2-bromophenoxy propane, o-bromoisopropoxybenzene, 2-bromoisopropoxybenzene, 1-bromo-2-1-methylethoxy benzene, 2-bromophenyl isopropyl ether, pubchem3828
PubChem CID 21925427
IUPAC Name 1-bromo-2-propan-2-yloxybenzene
SMILES CC(C)OC1=CC=CC=C1Br

Specifications

Density 1.34
Boiling Point 73°C (2 mmHg)
Odor Aromatic
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-isopropoxybenzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only