missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-chloro-3-fluorobenzene 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 209.44 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystals |
| Chemical Name or Material | 1-Bromo-2-chloro-3-fluorobenzene |
Safety and Handling
| TSCA | No |