missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dimethoxybenzene 97.0+%, TCI America™

Catalog No. B14815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14815G 5 g
B148125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14815G Supplier TCI America Supplier No. B14815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17715-69-4
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00009844
InChI Key NIUZVSQOXJIHBL-UHFFFAOYSA-N
Synonym 2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
PubChem CID 87266
IUPAC Name 1-bromo-2,4-dimethoxybenzene
SMILES COC1=CC=C(Br)C(OC)=C1

Specifications

Color White-Yellow
Boiling Point 132°C
Quantity 5 g
Formula Weight 217.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-2,4-dimethoxybenzene