missing translation for 'onlineSavingsMsg'
Learn More

1-(Boc-amino)cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH6199203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6199203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6199203 Supplier Thermo Scientific Chemicals Supplier No. H6199203

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88950-64-5
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.22
MDL Number MFCD00083257
InChI Key DSKCOVBHIFAJRI-UHFFFAOYSA-N
Synonym 1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino
PubChem CID 688396
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopropane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CC1)C(O)=O

Specifications

Melting Point 118°C (decomposition)
Beilstein 4430628
Quantity 1 g
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material 1-(Boc-amino)cyclopropanecarboxylic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only