missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals

Catalog No. AAH5795103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5795103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5795103 Supplier Thermo Scientific Chemicals Supplier No. H5795103
Only null left

1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 454703-20-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD08752496
InChI Key CHRBSEYIEDTNSC-UHFFFAOYSA-N
Synonym tert-butyl 3-methylamino azetidine-1-carboxylate, 1-boc-3-methylamino azetidine, 1-n-boc-3-methylamino azetidine, 1-azetidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-methylamino azetidine, 1-boc-3-methylaminoazetidine, 1-azetidinecarboxylicacid, 3-methylamino-, 1,1-dimethylethyl ester, 1-boc-3-methylaminoazatidine, 1-boc-3-methyl amino azatidine hydrochloride, 3-methylamino-1-azetidinecarboxylic acid tert-butyl ester
PubChem CID 23153172
IUPAC Name tert-butyl 3-(methylamino)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)NC

Specifications

Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 186.25
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material 1-Boc-3-(methylamino)azetidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only