missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-3-phenylthiourea 98.0+%, TCI America™
Supplier: TCI America B49975G
Specifications
| 1-Benzyl-3-phenylthiourea | |
| 155°C | |
| C14H14N2S | |
| 5 g | |
| NXCBDDGSOXJEFZ-UHFFFAOYSA-N | |
| 1-benzyl-3-phenylthiourea | |
| 668300 | |
| ≥98.0% (HPLC,N) |
| 726-25-0 | |
| White | |
| MFCD00022120 | |
| 2811 | |
| C1=CC=C(C=C1)CNC(=S)NC2=CC=CC=C2 | |
| 242.34 | |
| 242.34 | |
| Crystalline Powder |
Chemical Identifiers
| 726-25-0 | |
| 242.34 | |
| NXCBDDGSOXJEFZ-UHFFFAOYSA-N | |
| 1-benzyl-3-phenylthiourea |
| C14H14N2S | |
| MFCD00022120 | |
| 668300 | |
| C1=CC=C(C=C1)CNC(=S)NC2=CC=CC=C2 |
Safety and Handling
TSCA : No