missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-phenylthiourea 98.0+%, TCI America™

Catalog No. p-7123889
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B49971G 1 g
B49975G 5 g
2 options

Catalog No. B49971G

Supplier: TCI America B49971G

Only null left

Chemical Identifiers

CAS 726-25-0
Molecular Formula C14H14N2S
Molecular Weight (g/mol) 242.34
MDL Number MFCD00022120
InChI Key NXCBDDGSOXJEFZ-UHFFFAOYSA-N
PubChem CID 668300
IUPAC Name 1-benzyl-3-phenylthiourea
SMILES C1=CC=C(C=C1)CNC(=S)NC2=CC=CC=C2

Specifications

Melting Point 155°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 242.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-3-phenylthiourea