accessibility menu, dialog, popup

UserName

CAS RN 726-25-0

IUPAC Name: 3-benzyl-1-phenylthiourea
Synonyms: 3-benzyl-1-phenylthiourea
Molecular Weight (g/mol): 242.34
Molecular Formula: C14H14N2S
InChi Key: NXCBDDGSOXJEFZ-UHFFFAOYSA-N
SMILES: S=C(NCC1=CC=CC=C1)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results