missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-(methylamino)pyrrolidine 96.0+%, TCI America™

Catalog No. B15585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15585G 5 g
B155825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15585G Supplier TCI America Supplier No. B15585G

Chemical Identifiers

CAS 96568-35-3
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00191309,MFCD00191310,MFCD00191308
InChI Key UEAYAIWNQQWSBK-UHFFFAOYNA-N
PubChem CID 13787478
IUPAC Name 1-benzyl-N-methylpyrrolidin-3-amine
SMILES CNC1CCN(CC2=CC=CC=C2)C1

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
UN Number 2735
Formula Weight 190.29
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-(methylamino)pyrrolidine