missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC12204EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12204EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12204EA Supplier Thermo Scientific Chemicals Supplier No. CC12204EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3541-37-5
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD01075041
InChI Key NXSVNPSWARVMAY-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
PubChem CID 736500
IUPAC Name 1-benzothiophene-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 10 g
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-2-carbaldehyde
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation.