missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyrene, 97%

Catalog No. AC104530050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104530050 5 g Glass bottle
AC104530010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104530050 Supplier Thermo Scientific Chemicals Supplier No. 104530050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1606-67-3 | C16H11N | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1606-67-3
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.27
MDL Number MFCD00004140
InChI Key YZVWKHVRBDQPMQ-UHFFFAOYSA-N
Synonym 1-aminopyrene, 1-pyrenamine, 3-aminopyrene, pyrene, amino, pyren-1-ylamine, aminopyrene, pyrenamine, alpha-aminopyrene, unii-luw9eo1681, ccris 760
PubChem CID 15352
IUPAC Name pyren-1-amine
SMILES NC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 12,1341
Formula Weight 217.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Aminopyrene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-521-3
RTECSNumber UR2275000
TSCA TSCA

RUO – Research Use Only