missing translation for 'onlineSavingsMsg'
Learn More

1-Aminobenzotriazole, 97%

Catalog No. AAJ63610MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63610MD 250 mg
AAJ6361003 1 g
AAJ6361006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63610MD Supplier Thermo Scientific Chemicals Supplier No. J63610MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suicide inhibitor of cytochrome P450 and chloroperoxidase | CAS: 1614-12-6 | C6H6N4 | 134.142 g/mol

1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

Solubility
Soluble in chloroform, DMSO, ethanol or water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 1614-12-6
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.142
MDL Number MFCD00132902
InChI Key JCXKHYLLVKZPKE-UHFFFAOYSA-N
Synonym 1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
PubChem CID 1367
IUPAC Name benzotriazol-1-amine
SMILES C1=CC=C2C(=C1)N=NN2N

Specifications

Melting Point 81°C to 85°C
Color Brown to White
Quantity 250 mg
Beilstein 607843
Solubility Information Soluble in chloroform, DMSO, ethanol or water
Formula Weight 134.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Aminobenzotriazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DM1235000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only