missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3,3-diethoxypropane, 97%

Catalog No. AC269850250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269850250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC269850250 Supplier Thermo Scientific Chemicals Supplier No. 269850250
Only null left

CAS: 41365-75-7 | C7H17NO2 | 147.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41365-75-7
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.22
InChI Key PXXMSHBZYAOHBD-UHFFFAOYSA-N
Synonym 1-amino-3,3-diethoxypropane, 3,3-diethoxypropylamine, 3-aminopropionaldehyde diethylacetal, 1-propanamine, 3,3-diethoxy, 3-aminopropionaldehyde diethyl acetal, propionaldehyde, 3-amino-, diethyl acetal, zlchem 1268, acmc-1apws, 3,3-diethoxy-propylamine, 3,3-diethoxypropyl amine
PubChem CID 521029
IUPAC Name 3,3-diethoxypropan-1-amine
SMILES CCOC(CCN)OCC

Specifications

Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 72°C (16.0 hPa)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4233 to 1.4253
Linear Formula (C2H5O)2CHCH2CH2NH2
Quantity 25 g
Specific Gravity 0.91
Solubility Information Solubility in water: miscible, hydrolysis. Other solubilities: miscible with alcohol and petroleum ether
Formula Weight 147.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Amino-3, 3-diethoxypropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only