missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Amino-2-propanol, 94%, remainder 2-amino-1-propanol

Catalog No. AAL1534330
Change view
Click to view available options
Quantity:
250 g
1000 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1534330 250 g
AAL153430B 1000 g
AAL153430E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1534330 Supplier Thermo Scientific Chemicals Supplier No. L1534330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-96-6 | C3H9NO | 75.11 g/mol

Amino-2-propanol is used to study the effect of temperature and water content on the molecular structure and hydrogen bonding of amino-2-propanol, which has been examined by Fourier transform near-infrared (FT-NIR) spectroscopy. Also used in used in the synthesis of protein kinase CK2 inhibitors used in the treatment of neoplasia and other infective diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino-2-propanol is used to study the effect of temperature and water content on the molecular structure and hydrogen bonding of amino-2-propanol, which has been examined by Fourier transform near-infrared (FT-NIR) spectroscopy. Also used in used in the synthesis of protein kinase CK2 inhibitors used in the treatment of neoplasia and other infective diseases.

Solubility
Fully miscible with water.

Notes
Air Sensitive and Hygroscopic, store away form air and water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 78-96-6
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008139
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine
PubChem CID 4
ChEBI CHEBI:19030
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Density 0.959
Boiling Point 160°C
Flash Point 71°C (159°F)
Refractive Index 1.448
Quantity 250 g
UN Number UN2735
Beilstein 605275
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible with water.
Formula Weight 75.11
Percent Purity 94%
Assay remainder 2-amino-1-propanol
Chemical Name or Material (+/-)-1-Amino-2-propanol
Show More Show Less
Hazard Category H227-H312-H314-H335
Hazard Statement GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 201-162-7
RTECSNumber UA5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only