missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methyl-2-propanol 98.0+%, TCI America™

Catalog No. A24425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24425G 5 g
A24421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24425G Supplier TCI America Supplier No. A24425G

Chemical Identifiers

CAS 2854-16-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD01697283
InChI Key LXQMHOKEXZETKB-UHFFFAOYSA-N
Synonym 1,1-Dimethylethanolamine
PubChem CID 95102
IUPAC Name 1-amino-2-methylpropan-2-ol
SMILES CC(C)(O)CN

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 g
UN Number 1993
Formula Weight 89.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Amino-2-methyl-2-propanol
RTECSNumber UA6125000
TSCA No