missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-2-pyrrolidone 98.0+%, TCI America™

Numéro de catalogue. A14185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A14185G 5 g
A141825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A14185G Fournisseur TCI America Code fournisseur A14185G
Il en reste null

Identifiants chimiques

CAS 932-17-2
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00014101
InChI Key YLHUPYSUKYAIBW-UHFFFAOYSA-N
PubChem CID 70266
IUPAC Name 1-acetylpyrrolidin-2-one
SMILES CC(=O)N1CCCC1=O

Spécifications

Boiling Point 231°C
Quantity 5 g
Formula Weight 127.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetyl-2-pyrrolidone
RTECSNumber UY5717000
TSCA No